Order Entry
Singapore
ContactUsLinkComponent
Acetic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Acetic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates
undefined
Acetic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8%
  • Evaporation residue:
    Max. 0.0001%
  • Water:
    Max. 0.1%
  • Calcium (0.1% Aqueous solution):
    Max. 0.2 ppm
  • Potassium (0.1% Aqueous solution):
    Max. 0.2 ppm
  • Magnesium (0.1% Aqueous solution):
    Max. 0.2 ppm
  • Sodium (0.1% Aqueous solution):
    Max. 0.5 ppm
  • Transmittance (255 nm):
    Min 10%
  • Transmittance (260 nm):
    Min 50%
  • Transmittance (270 nm):
    Min 80%
  • Transmittance (280 nm):
    Min 95%
  • Transmittance (300 nm):
    Min 99%
  • Suitability for LC-MS (0.1% aqueous solution):
    Passes Test

Specification Test Results