Order Entry
Singapore
ContactUsLinkComponent
Connecting adapters, Claisen type, PYREX®
Connecting adapters, Claisen type, PYREX®
Catalog # CORN9040-24
Supplier:  Corning
undefined
Connecting adapters, Claisen type, PYREX®
Catalog # CORN9040-24
Supplier:  Corning
Specifications
  • Cone size:
    24/40 NS
  • Socket size:
    24/40 NS
  • Pk:
    6
Specifications
About this item
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These Claisen adapters have 24/40 outer standard taper joints at the upper end and on the parallel sidearm and an inner 24/40 standard taper joint at the lower end.