Order Entry
Singapore
ContactUsLinkComponent
 
Potassium citrate monohydrate 99.0-101.0%, granular, BAKER ANALYZED®, J.T.Baker®
  BAKR3066-01
 :  Avantor
 :  
undefined
Potassium citrate monohydrate 99.0-101.0%, granular, BAKER ANALYZED®, J.T.Baker®
  BAKR3066-01
 :  Avantor
 :  

 

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0 %
  • Assay (HOC(COOK)(CH₂COOK)₂·H₂O):
    99.0 - 101.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    7.0 - 9.0
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Calcium (Ca):
    ≤0.005 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Sodium (Na):
    ≤0.2 %
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

 

 

 :
 :13,07706
 :230 °C
 :1,98 g/cm³ (20 °C)
 :324.41 g/mol
 :212-755-5
 :6100-05-6
 :HOC(COOK)(CH₂COOK)₂·H₂O
 :MFCD00150442