Order Entry
Singapore
Orders LinkContactUsLinkComponent
Connecting adapter, PYREX®
24/40 NS, 24/40 NS
  CORN8930-24
 :   Corning
Connecting adapter, PYREX®
  CORN8930-24
 :  Corning
Connecting adapter, PYREX®
Connecting adapter, PYREX®
24/40 NS, 24/40 NS
$160.00
1 * 1 items
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These connecting distilling tube adapters have full-length inner 24/40 standard taper joints of same size. The connecting arm is approximately at a 75° angle from vertical and is 200 mm long.
  • Cone size:
    24/40 NS
  • Socket size:
    24/40 NS