Order Entry
Singapore
ContactUsLinkComponent
Three-Way Distilling Connecting Adapter with Two 24/40 Standard Taper Joints, PYREX®
Three-Way Distilling Connecting Adapter with Two 24/40 Standard Taper Joints, PYREX®
Catalog # CORN9060-24
Supplier:  Corning
undefined
Three-Way Distilling Connecting Adapter with Two 24/40 Standard Taper Joints, PYREX®
Catalog # CORN9060-24
Supplier:  Corning
Specifications
  • Cone size:
    24/40 NS
  • Socket size:
    10/30 NS
  • Pk:
    6
Specifications
About this item
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These distilling connecting adapters have a sidearm at an approximately 75° angle from the lower part of the vertical tube. The joint on the sidearm is at an angle approximately 105° from the sidearm. The sidearm and lower tube have standard taper inner joints. The upper tube is equipped with an outer standard taper joint for a thermometer.