Order Entry
Singapore
ContactUsLinkComponent
Adapters, with tubing connector, PYREX®
  CORN9420-24
 :  Corning
undefined
Adapters, with tubing connector, PYREX®
  CORN9420-24
 :  Corning
  • Cone size:
    24/40 NS
  • Socket size:
    24/40 NS
  • Pk:
    6
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
These distilling suction tube adapters convert ordinary boiling flasks with 24/40 standard taper jointed necks to vacuum-type standard taper receivers. They have a 24/40 standard taper outer joint at the top and a 24/40 standard taper inner joint at the bottom. The sidearm is approximately 10 mm Ø by 25 mm long.