Order Entry
Singapore
Orders LinkContactUsLinkComponent
Drying trays, PYREX®
Drying trays, PYREX®
Supplier:   Corning
Drying trays, PYREX®
Supplier:  Corning
Certificates
Drying trays, PYREX®
Drying trays, PYREX®
3 Options Available Below
Product Details & Documents
These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.
  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean
They can be used with gel electrophoresis or paper chromatography.
Caution: For oven use only. Do not use over direct heat.
Documents
Product Family Options
Product Information