Order Entry
Singapore
Orders LinkContactUsLinkComponent
Jars, cylindrical, PYREX®
Jars, cylindrical, PYREX®
Supplier:   Corning
Jars, cylindrical, PYREX®
Supplier:  Corning
Certificates
Jars, cylindrical, PYREX®
Jars, cylindrical, PYREX®
2 Options Available Below
Product Details & Documents
PYREX® glass.
Ideal for use as animal jars or for other purposes where resistance to hot air and steam sterilisation is of importance. The open ends are finished with a fire polished bead to increase mechanical strength.
Caution: Note: Do not use with heat, vacuum, or pressure applications.
Documents
Product Family Options
Product Information