Adapters, glass, reduction, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
[ST] outer joint at top
Larger [ST] inner joint at bottom.
Adapters feature a [ST] outer joint at the top and a larger [ST] inner joint at the bottom.