Micro burettes, straight stopcocks, class A, PYREX®
Supplier: Corning
Jump to:
About this item
Micro burettes, Class A, PYREX® borosilicate glass.
- Precision bore tubing
- Funnel top
- Tapered tips for smooth flow
- Stopcocks turn freely yet give positive control
Designed for precise analytical methods in microchemistry.
Certifications: The capacity tolerance on these burettes is established by ASTM E1189 and are calibrated in accordance with ASTM E542.
2 Options Available Below
Product Details & Documents
Micro burettes, Class A, PYREX® borosilicate glass.
- Precision bore tubing
- Funnel top
- Tapered tips for smooth flow
- Stopcocks turn freely yet give positive control
Designed for precise analytical methods in microchemistry.
Certifications: The capacity tolerance on these burettes is established by ASTM E1189 and are calibrated in accordance with ASTM E542.