Measuring cylinders, tall form, borosilicate glass, PYREX® VISTA™
Supplier: Corning
Certificates
About this item
Made of PYREX® glass, blue graduations.
- Calibrated to contain (TC)
- Tolerances are ±5% of total at any point
- 10 ml cylinder has a funnel top
They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Cylinders can be conveniently stacked when plastic bases are removed.
Delivery: Supplied with bumper guards for cylinders of capacity 25 to 100 ml. Snap caps are supplied with all sizes.