Volumetric flasks, class A, glass, clear, snap cap, PYREX® Lifetime Red™
: Corning
PYREX® borosilicate glass, clear, snap cap.
These flasks have the capacity mark etched as a fine line through a narrow red band into clear glass.
: ASTM E-694, E-542, E-288.