Order Entry
Singapore
Orders LinkContactUsLinkComponent
Funnels, stemless, PYREX®
 :   Corning
Funnels, stemless, PYREX®
 :  Corning
Funnels, stemless, PYREX®
Funnels, stemless, PYREX®
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These stemless funnels are of molded construction with bottoms ground flat and have a 6 mm opening. For use in the sugar industries.
Product Family Options
  • Length
    Int.Ø top
  • 60 mm
    75 mm
    1 * 48 items
    1 * 48 items - $1,010.00
    $1,010.00
    1 * 48 items
       $1,010.00
  • 82 mm
    100 mm
    1 * 1 items
    1 * 1 items - $34.20
    $34.20
    1 * 1 items
       $34.20