Volumetric flasks, class A, low actinic, glass stopper, PYREX®
: Corning
PYREX®, borosilicate glass, amber, low actinic, with tapered ground stopper.
- Individually serialised
- Markings are sharp and permanent
- Low actinic glass provide protection for material sensitive to light
Each flask is individually serialised and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. The protective color is an integral part of the flask, which retains its mechanical strength, chemical stability and thermal resistance.
: Calibrated to meet ASTM specifications E-694, E-542, and E-288