Oral gavage needles, disposable
Supplier: CADENCE SCIENCE
Certificates
About this item
Disposable sterile feeding needles are for critical applications.
- All needles feature a plastic luer hub and a soft silicone tip
- Malleable to any angle
- Non-toxic and non-pyrogenic
- Available with malleable stainless steel or flexible plastic (PTFE) tubing