Single metric scale graduated cylinders, to contain, PYREX® Lifetime Red™
: Corning
Calibrated ‘To Contain’ the scale and figures are durable white enamel, which stand out clearly against the red color-band on these lifetime red cylinders.
The 10 ml size has a funnel top. The 100 ml size is made in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86 and D-216.
: Bumper guards are supplied with 100 ml through 2000 ml inclusive.