Flasks, round bottom, glass, clear, with short neck, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, clear, with short neck.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These flask mouth tooled for uniform rubber stopper fit. The normal working temperature is 230 °C.