Boiling flasks, Florence, flat bottom, long neck, with standard taper joint, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, long neck.
- High thermal shock resistance
- Provide contamination free results
- High chemical durability
These flasks have full length outer standard tapered 24/40 joint. The normal working temperature is 230 °C.