Order Entry
Singapore
Orders LinkContactUsLinkComponent
Extraction thimbles, PYREX®
Extraction thimbles, PYREX®
Supplier:   Corning
Extraction thimbles, PYREX®
Supplier:  Corning
Certificates
Extraction thimbles, PYREX®
Extraction thimbles, PYREX®
5 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, with fritted disc.
  • Resistant to chemical attack
  • Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Documents
Product Family Options
Product Information
  • Ø ext.×H
    Availability
    Price
  • CORN33950-SC
    Coarse porosity fritted thimble, 40-60 μm
    Coarse porosity fritted thimble, 40-60 μm
    25×85 mm
    Log in for availability
    Log in for availability
    1 * 9 items
    1 * 9 items - $867.00
    $867.00
    1 * 9 items
       $867.00
  • CORN33950-MC
    Coarse porosity fritted thimble, 40-60 μm
    Coarse porosity fritted thimble, 40-60 μm
    35×90 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $108.00
    $108.00
    1 * 1 items
       $108.00
  • CORN33950-LC
    Coarse porosity fritted thimble, 40-60 μm
    Coarse porosity fritted thimble, 40-60 μm
    45×130 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $136.00
    $136.00
    1 * 1 items
       $136.00
  • CORN33950-MEC
    Extra coarse porosity fritted thimble, 170-220 μm
    Extra coarse porosity fritted thimble, 170-220 μm
    35×90 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $111.00
    $111.00
    1 * 1 items
       $111.00
  • CORN33950-LEC
    Extra coarse porosity fritted thimble, 170-220 μm
    Extra coarse porosity fritted thimble, 170-220 μm
    45×130 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $140.00
    $140.00
    1 * 1 items
       $140.00