Order Entry
Singapore
ContactUsLinkComponent
Measuring cylinder, tall form, borosilicate glass, percent scale graduated, PYREX®
Measuring cylinder, tall form, borosilicate glass, percent scale graduated, PYREX®
Catalog # CORN3065-200
Supplier:  Corning
Measuring cylinder, tall form, borosilicate glass, percent scale graduated, PYREX®
Catalog # CORN3065-200
Supplier:  Corning

Specifications

  • Capacity:
    200 ml
  • Height:
    254 mm
  • Pk:
    1

Specifications

About this item

Borosilicate glass.

  • Calibrated to contain TC in percent
  • Graduations are in durable white enamel
  • Used to measure the distillate in ASTM D86
Delivery: Supplied with bumper guards.