Volumetric flasks, class A, white markings, PYREX®
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, clear, with LDPE stoppers.
- Graduation line is sharp and permanent
- White marks and inscriptions are easy to read
Volumetric flasks is made with increased strength through machine blown bodies. Heavy tubing rings and heavy beaded seal is tooled for polyethylene tubing.