Order Entry
Singapore
Orders LinkContactUsLinkComponent
Hydrometer cylinders, PYREX®
Supplier:   Corning
Hydrometer cylinders, PYREX®
Supplier:  Corning
Certificates
Hydrometer cylinders, PYREX®
Hydrometer cylinders, PYREX®
5 Options Available Below
Product Details & Documents
These PYREX® glas hydrometer cylinders have a large hexagonal base sealed to the cylinder body to increase stability. Breakage is minimised by the heavy wall construction.
  • Excellent chemical resistance
  • Good temperature resistance
Meets ASTM specifications D-287 and E-100.
Product Family Options
Product Information
  • Capacity
    Int.Ø
    I-height
    Availability
    Price
  • 100 ml
    29 mm
    254 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $40.00
    $40.00
    1 * 1 items
       $40.00
    Specifications:
    More
    Specifications
    Cat. No.
    CORN2962-100
  • 250 ml
    39 mm
    340 mm
    Log in for availability
    Log in for availability
    1 * 18 items
    1 * 18 items - $940.00
    $940.00
    1 * 18 items
       $940.00
    Specifications:
    More
    Specifications
    Cat. No.
    CORN2962-250
  • 300 ml
    39 mm
    381 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $58.00
    $58.00
    1 * 1 items
       $58.00
    Specifications:
    More
    Specifications
    Cat. No.
    CORN2962-300
  • 500 ml
    50 mm
    390 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $69.50
    $69.50
    1 * 1 items
       $69.50
    Specifications:
    More
    Specifications
    Cat. No.
    CORN2962-500
  • 1000 ml
    64 mm
    465 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $101.00
    $101.00
    1 * 1 items
       $101.00
    Specifications:
    More
    Specifications
    Cat. No.
    CORN2962-1L