Order Entry
Singapore
ContactUsLinkComponent
Beakers, low form, Griffin, PYREX® VISTA™
Beakers, low form, Griffin, PYREX® VISTA™
Catalog # CORN70000-10
Supplier:  Corning
Supplier #: 70000-10
undefined
Beakers, low form, Griffin, PYREX® VISTA™
Catalog # CORN70000-10
Supplier:  Corning
Supplier #: 70000-10
Log in for availability
Select an Option
25 mm
Select an Option
10* ml
Select an Option
33 mm
Select an Option
01
Select an Option
24
About this item
These beakers with spout are made of Type I Class A low expansion borosilicate glass, per ASTM E438.
  • High mechanical strength
  • Resistance to thermal shock
Beakers have blue graduations and an extra large blue marking spot. The 250 through 1000 ml beakers have a double graduated metric scale to indicate approximate content.
*10 ml beaker is not graduated.
Certifications: ASTM E-960