Chemistry kit, PYREX®
Supplier: Corning
Certificates
About this item
Chemistry kit is designed for most common organic chemistry experiments.
It comprises 15 components with 19/22 standard tapered joints including single neck flasks, a 3 neck flask suitable for steam distillation, adapters and condensers. The components are held in foam within a polyethylene case.