Aspirator bottles with tubing outlet, PYREX®
Supplier: Corning
Certificates
About this item
PYREX®, borosilicate glass, without stoppers.
- Excellent chemical resistance
- High thermal resistance, autoclavable
- Minimal thermal expansion giving relatively high resistance to temperature changes
Bottles with tubular sidearm outlet facilitates attachment of flexible tubing and it withstands repeated wet or dry autoclaving. It is designed for use as a delivery or storage container for solutions.