Glove liners, HyFlex® 48-100 / 48-101
Supplier: Ansell
Certificates
About this item
These knitted nylon gloves with PU coated palm are designed for light applications that require good levels of comfort and abrasion resistance.
- Optimum protection for light tasks, high level of comfort
- Good freedom of movement for precise handling of small objects
- Elasticated nylon liner for exceptionally close fit
- Excellent alternative to light leather gloves
- Available in white or black
Ideal for the precise handling of small objects in dry or light oily environments.
Colour: White
EN 388 (003X), Cat. II
Certifications: EN 388:2016 (4131A); Cat. II
Specifications
- Catalog No:
- ANSE48-100/7
- ANSE48-100/8
- ANSE48-101/8
- ANSE48-100/10
- ANSE48-101/9
- ANSE48-100/11
- ANSE48-100/6
- ANSE48-101/10
- ANSE48-101/11
- ANSE48-101/6
- ANSE48-101/7
- ANSE48-100/9
- Model:
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- 48-100
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)
- EN 388:2016 (4131A)