Bovine RNase I 'A' (from Pancreas)
Catalog # USBM27032301100MG
Supplier: USB MOLECULAR BIOLOGY REAGENTS
CAS Number:
Specifications
- Pk:100 mg
- Source:Pancreas
- Species:Bovine
- Environmentally Preferable:
- Enzyme name:RNase I 'A'
Specifications
About this item
Source: Bovine pancreas.