Order Entry
Singapore
ContactUsLinkComponent
VWR® CERTICLEAN®, Cleanroom Gloves, Latex
undefined
VWR® CERTICLEAN®, Cleanroom Gloves, Latex
  • Catalog No:
  • VWRU40101-152
  • VWRU40101-150
  • VWRU40101-148
  • VWRU40101-146
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Finger thickness:
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • Palm thickness:
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm