VWR®, Hygiene Masks, Advanced Protection
VWR® Advanced Protection face masks feature a three-ply construction for excellent particle and bacterial filtration efficiency. Available in two styles, with spandex ear loops or PP ties ultrasonically welded to maintain softness and protect against particulates.
- Latex-free
- Guards against bacteria and particulates
- Highly breathable and fluid-resistant
- Malleable 12 cm noseband ensures custom, secure seal
- Soft and hypoallergenic
: Masks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards. Products are validated through independent laboratory testing.
: 50 masks per pack, 10 packs per case.