Dialamatic microdispensers
Dialamatic microdispenser, adjustable, 2 - 10 µl, ±1% , 1 μl
Catalog # DRUM3-000-210
Supplier: DRUMMOND
Jump to:
Product Details & Documents
Dialamatic microdispensers with Borosilicate glass bores with stainless steel plungers (PTFE tips on all plungers 25 μl or larger).
- Adjustable, direct reading plunger sets volume
- Delivery scale engraved on plunger shaft
- Fractional deliveries possible with premarked indexes
- Volumetric accuracy of ±1% (at full volume)
Specifications
- Capacity:2 - 10 µl
- Description:Dialamatic microdispenser, adjustable
- Seq:01
- Accuracy:±1%
- Division:1 μl
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement bore for microdispenser, black, 20 µlReplacement bore for microdispenser, black, 20 µlDRUM3-000-320-GLog in for availability1 * 100 items - $88.50
Product Information
- Catalog #AvailabilityPrice
- Replacement bore for microdispenser, orange, 10 µlReplacement bore for microdispenser, orange, 10 µlDRUM3-000-210-GLog in for availability1 * 100 items - $42.10