Reagent reservoirs, disposable
Supplier: VISTALAB TECHNOLOGIES
Certificates
About this item
These reservoirs made of PS feature a trough-within-a-trough design to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
- Pour-off spouts at every corner
- Wider base for stability
- Free of animal products
- Sterile reservoirs are certified RNase/DNase free and non pyrogenic