Boot covers, Tyvek® IsoClean, model IC 458, sterile
Supplier: DuPont
Certificates
About this item
These sterile boot covers are composed of Tyvek® fabric (flash spun high density PE). Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
- Ultra low linting fabric
- Fixation ankle ties, tunnelled elastics at ankles
- Gripper™ soles provide enhanced skid resistance and durability
- Clean processed and gamma sterilised
- Bound seams: Tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
Ideal for use in biotech, pharmaceutical and medical device manufacturing, in electronics industries, as well as in other critical or controlled environments.
Height: 457 mm
Packaging: 100 per box, individually packed.
Specifications
- Catalog No:
- DUPOIC458BWHM0100
- DUPOIC458BWHXL0100
- DUPOIC458BWHS0100
- DUPOIC458BWHL0100
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- Garment application:
- Cleanroom, Pharmaceutical
- Cleanroom, Pharmaceutical
Specifications
Related Information
Cleanroom_Whitepaper_Advantages of IsoClean_EN_Final DUPONT™ TYVEK® ISOCLEAN®Sizing Chart Accessories
Application Notes
Controlled_Environments_Catalogue_2016_EN DUPONT™ TYVEK® ISOCLEAN® DUPONT™ TYVEK® ISOCLEAN® FAQ Poster Non-Sterile Cleanroom Gowning Procedure