Cut resistant gloves, ActivArmr® 80-658
: Ansell
This glove consists of a durable liner made of stainless steel/nylon/Kevlar®/glass fiber/cotton and a ¾ dipped natural rubber coating. Thanks to its triple-layer construction (steel/glass fibre/Kevlar®) it provides extreme resistance to cuts, splinters and burrs.
- Extra-long cuff offers extended safety for the forearm
- Glove is ¾ dipped and liquid repellent, shielding the skin from unpleasant liquid solutions
- Good heat contact resistance for handling of warm objects
- Natural rubber crinkle finish provides a firm grip
- Excellent washability, with no decrease in comfort or fading
Ideal for handling heavy loads, glass sheets and sharp-edged objects, recycling, sorting (glass, cans, metal pieces) and cutting of dry, painted or galvanised pieces.
: EN 388:2016 (XX43E), EN 407 (X2XXXX); Cat. III
- Catalog No:
- ANSE80-658/8
- ANSE80-658/9
- ANSE80-658/10
- ANSE80-658/11
- ANSE80-658/7
- ANSE80-658/6
- Model:
- 80-658
- 80-658
- 80-658
- 80-658
- 80-658
- 80-658
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Kevlar®/cotton
- Kevlar®/cotton
- Kevlar®/cotton
- Kevlar®/cotton
- Kevlar®/cotton
- Kevlar®/cotton
- Finish:
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- Gauge:
- 7
- 7
- 7
- 7
- 7
- 7
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (XX43E), EN 407 (X2XXXX)
- EN 388:2016 (XX43E), EN 407 (X2XXXX)
- EN 388:2016 (XX43E), EN 407 (X2XXXX)
- EN 388:2016 (XX43E), EN 407 (X2XXXX)
- EN 388:2016 (XX43E), EN 407 (X2XXXX)
- EN 388:2016 (XX43E), EN 407 (X2XXXX)