Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lithium chloride
Lithium chloride 200 mM in aqueous solution
Lithium chloride 200 mM in aqueous solution
Lithium chloride 200 mM in aqueous solution
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Lithium chloride 200 mM in aqueous solution
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7447-41-8
:
Formula:
LiCl
MDL Number:
MFCD00011078
:
42.39 g/mol
Storage Temperature:
Ambient
EINECS:
231-212-3
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ62644
Videos
Thermo Scientific Acroseal 25 Years
Documentation