

Methanol : Water (50:50)% (v/v), LabReady™, Solvent Blend for HPLC, for spectrophotometry, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
:67-56-1
Formula:CH₄O/H₂O
MDL Number:MFCD00004595
Merck Index:12,08153
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0,9264 g/cm³ (25 °C)
Flash Pt:29 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Water (v/v):49.0 - 51.0%
- Methanol (v/v):49.0 - 51.0%
- UV Cutoff:198 nm
- Residue:< 1 mg/L
- Color (APHA):< 10
- HPLC Gradient @ 220 nm:< 0.005 AU
- HPLC Gradient @ 254 nm:< 0.001 AU