


Acetonitrile : Water (75:25)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJAA800-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Pk Info:4 x 4l
Specifications
About this item
:
Boiling Pt:66,67 °C
Density:0,8478 g/cm³ (20 °C)
Flash Pt:8,89 °C
Storage Temperature:Ambient
EINECS:200-929-3
UN:1648
ADR:3,II
:GHS07
:GHS08
:GHS02
:75-05-8