Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-tert-Butyl-3-ethylbenzene
1-tert-Butyl-3-ethylbenzene ≥98%
1-tert-Butyl-3-ethylbenzene ≥98%
1-tert-Butyl-3-ethylbenzene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
1-tert-Butyl-3-ethylbenzene ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
14411-56-4
:
Formula:
C₁₂H₁₈
MDL Number:
MFCD00060845
:
162.27 g/mol
Boiling Pt:
205 °C
Density:
0,86
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB20325
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation