Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Oxazepane hydrochloride
1,4-Oxazepane hydrochloride ≥98%
1,4-Oxazepane hydrochloride ≥98%
Irritant
1,4-Oxazepane hydrochloride ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1,4-Oxazepane hydrochloride ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
178312-62-4
:
5638-60-8
Formula:
C₅H₁₂ClNO
MDL Number:
MFCD02683070
:
137.61 g/mol
Melting Pt:
158…164 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB21749
Videos
Thermo Scientific Acroseal 25 Years
Documentation