
Biphenyl-4-sulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1623-93-4
:
Formula:C₁₂H₉ClO₂S
MDL Number:MFCD00078658
:252.72 g/mol
Boiling Pt:200…205 °C (6 mmHg)
Melting Pt:112…117 °C
Storage Temperature:Ambient
EINECS:216-609-1
ADR:8,II
:GHS05