Order Entry
Singapore
ContactUsLinkComponent
3-Phenylaniline hydrochloride
Health
Irritant
3-Phenylaniline hydrochloride
Catalog # ALFAH51027.06
CAS Number:  
Health
Irritant
3-Phenylaniline hydrochloride
Catalog # ALFAH51027.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:2113-55-5
 :
Formula:C₁₂H₁₂ClN
MDL Number:MFCD00068358
 :205.69 g/mol
 :GHS08
 :GHS07
Storage Temperature:Ambient