Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-Methoxy-5-nitrophenyl)acetic acid
(2-Methoxy-5-nitrophenyl)acetic acid ≥97%
(2-Methoxy-5-nitrophenyl)acetic acid ≥97%
Irritant
(2-Methoxy-5-nitrophenyl)acetic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
(2-Methoxy-5-nitrophenyl)acetic acid ≥97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
51073-04-2
:
Formula:
C₉H₉NO₅
MDL Number:
MFCD02664832
:
211.17 g/mol
Melting Pt:
153…155 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAH34386
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation