

(R)-(-)-2-(Methoxymethyl)pyrrolidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84025-81-0
:
Formula:C₆H₁₃NO
MDL Number:MFCD00066219
:115.18 g/mol
Boiling Pt:61…62 °C (40 mmHg)
Density:0,933
Flash Pt:45 °C (113 °F)
Storage Temperature:Ambient
ADR:3,III
:GHS02
:GHS07