Order Entry
Singapore
ContactUsLinkComponent
3-(1H-Pyrazol-3-yl)pyridine 95%
Irritant
3-(1H-Pyrazol-3-yl)pyridine 95%
Catalog # ALFAH33201.03
CAS Number:  
Irritant
3-(1H-Pyrazol-3-yl)pyridine 95%
Catalog # ALFAH33201.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :145.16 g/mol
Melting Pt:57…60 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:45887-08-9
Formula:C₈H₇N₃
MDL Number:MFCD03407951