Methyl-2-amino-4-fluorobenzoate 97%
Catalog # ALFAH61749.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:169.15 g/mol
Storage Temperature:Ambient
CAS Number:2475-81-2
Formula:C₈H₈FNO₂
MDL Number:MFCD11054082