Breast biobsy container
SIMPM970-D5B-1
- Description:Breast biobsy container, 5 compartments
- Environmentally Preferable:
- Pk:10
Breast biobsy container for separation, imaging and transport of core needle specimens. Designed specifically for radiography, separation, imaging and transport of core needle breast biopsies.
- Leakproof seal, thanks to O-ring lid, allows for safe and easy transport of the specimens from collection to analysis
- Not formalin prefilled
Special absorbent liner keeps specimens moist (when saline solution is added) prior to radiography while helping to attenuate the x-ray beam. Four compartments are clearly identified (3, 6, 9 and 12) and the radiolucent numbers show up clearly on the radiograph.
Formalin may be added prior to transportation to pathology for analysis. An area for writing patient information is provided on the label.