Order Entry
Singapore
ContactUsLinkComponent
Shorr staining solution, Q Path® for microscopy
Flammable
Shorr staining solution, Q Path® for microscopy
Catalog # VWRQ10047013
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Shorr staining solution, Q Path® for microscopy
Catalog # VWRQ10047013
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Pouch
  • Pk:
    450 mL
  • Seq:
    2675

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Colour of solution:
    Purplish black
  • Density (20/4):
    0,89 - 0,914
  • pH (20°C):
    3.1 - 3.8
  • Absorbance (0.1 %):
    500 - 503 nm

Specification Test Results

About this item

 :
Storage Temperature:Ambient
UN:1170
ADR:3,II
 :GHS02
 :1