Order Entry
Singapore
ContactUsLinkComponent
Thymolphthalein 0,5 g/l in ethanol 90% (v/v)
 
Thymolphthalein 0,5 g/l in ethanol 90% (v/v)
  RICC8450-16
 :  
 
Thymolphthalein 0,5 g/l in ethanol 90% (v/v)
  RICC8450-16
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

 :
 :430.54 g/mol
 :13,09478
 :Ambient
 :204-729-7
 :1170
 :3,II
 :GHS02
 :125-20-2
 :C₂₈H₃₀O₄
 :MFCD00005909