Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')europium(III)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17631-68-4
:
Formula:C₃₀H₃₀EuF₂₁O₆
MDL Number:MFCD00064655
:1037.49 g/mol
Melting Pt:203…207 °C
Storage Temperature:Ambient
EINECS:241-616-1