

Water with 0,1% (v/v) acetic acid ≥70% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:67-63-0
:7732-18-5
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0,9982 g/cm3
Storage Temperature:Ambient
EINECS:200-661-7
UN:1987
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Acetic Acid (v/v):0.09 - 0.11%
- UV Cutoff:<193 nm
- Residue:< 1 mg/L
- Color (APHA):< 10
- HPLC Gradient:Passes test