Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromocresol green
Bromocresol green 0,1%
Bromocresol green 0,1%
MANA814920
:
MACHEREY-NAGEL
:
Bromocresol green 0,1%
MANA814920
:
MACHEREY-NAGEL
:
Pk:
1
Seq:
0001
:
:
698.02 g/mol
:
Ambient
:
200-972-8
:
1219
:
3,II
:
GHS02
:
GHS07
:
76-60-8
:
C₂₁H₁₄Br₄O₅S
:
MFCD00005874